Influenza A H5N1 (A/Hong Kong/483/1997) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
11689-V08H-100
103622-278EA 351.48 USD
103622-278
Influenza A H5N1 (A/Hong Kong/483/1997) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Gln 531) of the influenza A H5N1 hemagglutinin (A/Hong Kong/483/1997 (H5N1)) (AAC32099.1), (HA1+HA2, uncleaved, cleavage site mutated (RRRKKR-TETR) was expressed, fused with a C-terminal polyhistidine tag.