Influenza A H1N1 (A/Puerto Rico/8/1934) Recombinant Hemagglutinin Protein (HA1 Subunit) (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
11684-V08H1-100
103634-428EA 351.48 USD
103634-428
Influenza A H1N1 (A/Puerto Rico/8/1934) Recombinant Hemagglutinin Protein (HA1 Subunit) (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the N-terminal segment (Met 1-Arg 343) of the influenza hemagglutinin (A/Puerto Rico/8/1934 (H1N1)) (ABD77675.1), termed as HA1, was fused with a C-terminal polyhistidine tag.