Human Recombinant DMBT1/Muclin Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
11678-H08H-100 11678-H08H-50
103613-506EA 212.85 USD
103613-506 103622-228
Human Recombinant DMBT1/Muclin Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the N-terminal segment of human DMBT1 (NP_004397.2) (Met 1-Ser 220) was expressed, fused with a polyhistidine tag at the C-terminus.