Human Recombinant Lumican/LUM Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
11640-H08H-50 11640-H08H-100
103681-324EA 191.35 USD
103681-324 103622-200
Human Recombinant Lumican/LUM Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human LUM (NP_002336.1) (Met 1-Asn 338) was fused with a polyhistidine tag at the C-terminus.