Human Recombinant NRG1-beta 1 Protein (EGF Domain, Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
11609-H01H-100 11609-H01H2-50
103622-164EA 300.98 USD
103622-164 103622-166
Human Recombinant NRG1-beta 1 Protein (EGF Domain, Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the EGF-like domain (Thr 176-Lys 246) of human NRG1 isoform beta1 (Q02297-6) was fused with the Fc region of human IgG1 at the N-terminus.