Human Recombinant FAM19A4/TAFA4 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
11165-H01H-50 11165-H01H-100
103624-160EA 300.98 USD
103624-160 103681-176
Human Recombinant FAM19A4/TAFA4 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human FAM19A4 (Q96LR4) (Ser36-Arg140) was expressed with the Fc region of human IgG1 at the N-terminus.