Human Recombinant Galectin-9/LGALS9 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
11147-H01H-50 11147-H01H-200 11147-H01H-50
103681-058EA 199.98 USD
CA76456-524 103681-058 103621-960
Human Recombinant Galectin-9/LGALS9 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human LGALS9 (NP_002299.2) (Ala2-Thr323) was expressed with the Fc region of human IgG1 at the N-terminus.