Human Recombinant Glucokinase/GCK Protein (from E. coli)
Supplier: Sino Biological
11078-HNCE-50 11078-HNCE-50 11078-HNCE-20
103680-530EA 212.85 USD
103680-530 103680-532 CA76456-024
Human Recombinant Glucokinase/GCK Protein (from E. coli)
Proteins and Peptides
A DNA sequence encoding the human glucokinase isoform 1 (NP_000153.1) (Leu 2-Gln 465) was expressed, fused with two additional amino acids (Gly and Pro) at the N-terminus.