Cynomolgus Recombinant IL13 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
11057-C07H-1 11057-C07H-100
103624-132EA 351.48 USD
103624-132 76631-374
Cynomolgus Recombinant IL13 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mature form of Cynomolgus (Macaca fascicularis) IL-13 (ABG75889.1) (Ser 21-Asn 132) was fused with a polyhistidine tag at the N-terminus.