Human Recombinant TMEFF1/Tomoregulin-1 Protein (Fc Tag, ECD) (from HEK293 Cells)
Supplier: Sino Biological
11005-H02H-20 11005-H02H-100
103680-350EA 62.35 USD
103680-350 103621-716
Human Recombinant TMEFF1/Tomoregulin-1 Protein (Fc Tag, ECD) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human TMEFF1 (NP_003683.2) (Met1-Val330) was expressed with the Fc region of human IgG1 at the C-terminus.