Human Recombinant IL10/Interleukin-10 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10947-H07H-5 10947-H07H-20 10947-H07H-100
103677-660EA 83.85 USD
103677-660 103681-040 103621-684
Human Recombinant IL10/Interleukin-10 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human IL10 (NP_000563.1) (Ser 19-Asn 178) was expressed, with a N-terminal polyhistidine tag.