Human Recombinant CXCL1/MGSA/NAP-3 Protein (His & NusA Tag) (from E. coli)
Supplier: Sino Biological
10877-H24E-50 10877-H24E-50 10877-H24E-20
103680-798EA 116.1 USD
CA76456-270 103680-798 103680-800
Human Recombinant CXCL1/MGSA/NAP-3 Protein (His & NusA Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the mature form of human CXCL1 (P09341-1) (Ala 35-Asn 107) was fused with the polyhistidine-tagged NusA tag at the N-terminus.