Human Recombinant CD70/CD27L/TNFSF7 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10780-H01H-5 10780-H01H-100 10780-H01H-50
103621-512EA 300.98 USD
103621-512 103678-976 103681-076
Human Recombinant CD70/CD27L/TNFSF7 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of human CD70 (NP_001243.1) (Gln 39-Pro 193) was fused with the Fc region of human IgG1 at the N-terminus via a polypeptide linker.