Human Recombinant ULBP1/RAET1/N2DL1 Protein (His & Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10679-H03H-100
103621-390EA 300.98 USD
103621-390
Human Recombinant ULBP1/RAET1/N2DL1 Protein (His & Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mature form of human ULBP1 (NP_079494.1) (Met 1-Gly 216) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.