Human Recombinant IGFBP3/IBP3 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10430-H07H-1 10430-H07H-100 10430-H07H-1
103677-572EA 2017.98 USD
103677-572 103621-208 CA76453-602
Human Recombinant IGFBP3/IBP3 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mature form of human IGFBP3 (NP_000589.2) (Gly28-Lys291) was fused with a polyhistidine tag at the N-terminus.