Human Recombinant Leptin Receptor/LEPR/CD295 Protein (His & Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10322-H03H-100 10322-H03H-200
103620-972EA 320.35 USD
103620-972 103620-970
Human Recombinant Leptin Receptor/LEPR/CD295 Protein (His & Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Asp 839) of human leptin receptor precursor (NP_002294.2) was expressed with the C-terminal fused polyhistidine-tagged Fc region of human IgG1.