Human Recombinant CLEC7A/Dectin-1/CLECSF12 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10215-H01H-50 10215-H01H-100
103678-592EA 191.35 USD
103678-592 103624-020
Human Recombinant CLEC7A/Dectin-1/CLECSF12 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human Dectin1 (NP_072092.2) (Thr 66- Met201) was expressed with the Fc region of human IgG1 at the N-terminus.