Human Recombinant EphB6/EphB6 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10197-H02H-100 10197-H02H-200
103620-880EA 199.98 USD
103620-880 103620-882
Human Recombinant EphB6/EphB6 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
The extracellular domain (Met 1-Ser 579) of human EphB6 (NP_004436.1) precursor was fused with the Fc region of human IgG1 at the C-terminus.