Human Recombinant CD31/PECAM1 Protein (from HEK293 Cells)
Supplier: Sino Biological
10148-HCCH-50 10148-HCCH-100 10148-HCCH-10 10148-HCCH-100
103678-428EA 148.35 USD
CA103623-882 103678-428 103678-426 103623-882
Human Recombinant CD31/PECAM1 Protein (from HEK293 Cells)
Proteins and Peptides
The mature form of human CD31 (EAW94208.1) extracellular domain (Met1-Lys 601) with five additional amino acids (DDDDK) at the C-terminus was expressed and purified.