Human Recombinant IL-1RA/IL1RN Protein (from E. coli)
Supplier: Sino Biological
10123-HNAE-100 10123-HNAE-100 10123-HNAE-200
103623-694EA 191.35 USD
103623-694 103623-692 CA103623-692
Human Recombinant IL-1RA/IL1RN Protein (from E. coli)
Proteins and Peptides
A DNA sequence encoding the mature form of human IL1RA isoform1 (NP_776214.1) (Arg 26-Glu 177) was expressed.