Human Recombinant JAML/AMICA Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10120-H08H-100 10120-H08H-50 10120-H08H-100
103620-790EA 300.98 USD
CA103620-790 103620-790 103678-356
Human Recombinant JAML/AMICA Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human JAML (NP_001091996.1) extracellular domain (Met 1-Leu 275) was fused with a polyhistidine tag at the C-terminus.