Human Recombinant IL27/Interleukin-27 Protein (His Tag) (from CHO cells)
Supplier: Sino Biological
10076-H08S-5 10076-H08S-5
103678-942EA 109.08 USD
103678-942 CA103678-942
Human Recombinant IL27/Interleukin-27 Protein (His Tag) (from CHO cells)
Proteins and Peptides
A DNA sequence encoding the IL27 complex composed of IL27B subunit (NP_005746.2) (Met 1-Lys 229) and the N-terminally truncated IL27p28 (IL27A) subunit (NP_663634.2) (Phe 29-Pro 243) linked by an polypeptide linker was fused with a polyhistidine tag at the C-terminus.