Mouse Recombinant CD155/PVR Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
50259-M08H-100
103664-820EA 300.98 USD
103664-820
Mouse Recombinant CD155/PVR Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse CD155 (NP_081790.1) precursor (Met 1-Arg 345) was expressed with a C-terminal polyhistidine tag.