Human Recombinant KIT/c-KIT/CD117 Protein (aa 540-972, His & GST Tag) (from Baculovirus Insect cells)
Supplier: Sino Biological
11996-H20B1-50
103622-914EA 199.98 USD
103622-914
Human Recombinant KIT/c-KIT/CD117 Protein (aa 540-972, His & GST Tag) (from Baculovirus Insect cells)
Proteins and Peptides
A DNA sequence encoding the human KIT isoform 2 (P10721-2) cytoplasmic domain (Thr 540-Val 972) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.