Human Recombinant CD16b/FCGR3B Protein (NA2 allotype), Biotinylated (from E. coli)
Supplier: Sino Biological
11046-HNAE-B-100 11046-HNAE-B-20
103632-658EA 159.58 USD
103632-658 103634-052
Human Recombinant CD16b/FCGR3B Protein (NA2 allotype), Biotinylated (from E. coli)
Proteins and Peptides
A DNA sequence encoding the mature form of human CD16b (O75015.2) (Met 18-Gly 193) was expressed and purified. The expressed protein was biotinylated in vitro.