Human Recombinant Progranulin Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10826-H08H-100
103621-552EA 300.98 USD
103621-552
Human Recombinant Progranulin Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human Progranulin (NP_002078.1) (Met 1-Leu 593) with a C-terminal polyhistidine tag was expressed.