Human Recombinant CD38 Protein (His & FLAG Tag) (from HEK293 Cells)
Supplier: Sino Biological
10818-H32H-50 10818-H32H-50
103680-042EA 452.48 USD
103680-042 CA76455-574
Human Recombinant CD38 Protein (His & FLAG Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of human CD38 (NP_001766.2) (Val 43 – Ile 300) was fused with a polyhistidine tag and a flag tag at the N-terminus.