Human Recombinant Beta-amyloid 40/Beta-APP40 Protein (His & GST Tag) (from E. coli)
Supplier: Sino Biological
10703-H20E1-100
103632-628EA 351.48 USD
103632-628
Human Recombinant Beta-amyloid 40/Beta-APP40 Protein (His & GST Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the amino acids (Asp 672-Val 711) of human Amyloid beta A4 protein (APP770) (P05067-1), corresponding to the Beta-amyloid protein 40, was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.