Human Recombinant TIM-3/HAVCR2 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10390-H08H-100
103621-132EA 351.48 USD
103621-132
Human Recombinant TIM-3/HAVCR2 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human HAVCR2 ( AAL65157.1) (Met1-Arg200) was expressed with a polyhistidine tag at the C-terminus.