Human Recombinant CD112/Nectin-2/PVRL2 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10005-H02H-100 10005-H02H-100
103620-678EA 300.98 USD
103620-678 CA103620-678
Human Recombinant CD112/Nectin-2/PVRL2 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Leu 360) of human CD112 (NP_002847.1) was fused with the Fc region of human IgG1 at the C-terminus.