Rhesus Recombinant B7-H5/GI24/VISTA Protein (ECD, Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90801-K02H-200
103585-672EA 452.48 USD
103585-672
Rhesus Recombinant B7-H5/GI24/VISTA Protein (ECD, Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rhesus C10orf54 (NP_001177807.1) (Met1-Ala194) was expressed with the Fc region of human IgG1 at the C-terminus.