Cynomolgus Recombinant IGFBP5/IGFBP-5 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
90283-C08H-100
103585-566EA 351.48 USD
103585-566
Cynomolgus Recombinant IGFBP5/IGFBP-5 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus IGFBP5 (F1PEQ5) (Met1-Glu271) was expressed with a polyhistidine tag at the C-terminus.