Cynomolgus Recombinant FAS/CD95/APO-1/TNFRSF6 Protein (from HEK293 Cells)
Supplier: Sino Biological
90273-CCCH-100
103585-604EA 300.98 USD
103585-604
Cynomolgus Recombinant FAS/CD95/APO-1/TNFRSF6 Protein (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus FAS (F6V1W6) (Met1-Asp173) was expressed with six amino acids (LEVLFQ) at the C-terminus.