Cynomolgus Recombinant CD53 Protein (aa 107-181, Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90189-C04H-100
103585-220EA 351.48 USD
103585-220
Cynomolgus Recombinant CD53 Protein (aa 107-181, Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus CD53 (G7NW46) (Glu107-Asn181) was expressed with the Fc region of mouse IgG1 at the N-terminus.