Cynomolgus Recombinant IFNA13/Interferon alpha-13 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
90108-C02H-100 90108-C02H-50
103669-792EA 169.85 USD
103669-792 103585-324
Cynomolgus Recombinant IFNA13/Interferon alpha-13 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus IFNA13 (G7NFW4) (Met1-Glu190) was expressed with the Fc region of human IgG1 at the C-terminus.