Rhesus Recombinant AMY2A/Alpha-amylase Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
90012-C08H-100
103585-064EA 351.48 USD
103585-064
Rhesus Recombinant AMY2A/Alpha-amylase Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rhesus AMY2A (H9EX43) (Met1-Leu511) was expressed with a polyhistidine tag at the C-terminus.