Rat Recombinant UCHL3 Protein (His Tag) (from E. coli)
Supplier: Sino Biological
80087-R07E-50
103669-194EA 300.98 USD
103669-194
Rat Recombinant UCHL3 Protein (His Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the rat UCHL3 (Q91Y78) (Glu 2-Ala 230) was fused with a polyhistidine tag at the N-terminus.