Rat Recombinant HER2/ErbB2 Protein (from HEK293 Cells)
Supplier: Sino Biological
80079-RCCH-50
103669-178EA 351.48 USD
103669-178
Rat Recombinant HER2/ErbB2 Protein (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the rat ErbB2 (AAH61863.1) extracellular domain (Met 4-Thr 656) was expressed and purified, with seven additional amino acids (ALEVLFQ) at the C-terminus.