Rat Recombinant IL-1 beta/IL1B Protein (mature form) (from E. coli)
Supplier: Sino Biological
80023-RNAE-20 80023-RNAE-100
103669-246EA 179.78 USD
103669-246 103666-244
Rat Recombinant IL-1 beta/IL1B Protein (mature form) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the mature form of rat IL1B (Q63264-1) (Val 117-Ser 268) was expressed and purified.