Canine Recombinant IL11RA/IL-11RA/IL11Rα Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
70078-D08H-50 70078-D08H-100
103665-768EA 300.98 USD
103665-768 103669-098
Canine Recombinant IL11RA/IL-11RA/IL11Rα Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the canine IL11RA (XP_005626799.1) (Met1-Leu363) was expressed with a C-terminal polyhistidine tag.