Canine Recombinant ANGPTL1/Angiopoietin-like 1 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
70032-D01H-50
103668-438EA 300.98 USD
103668-438
Canine Recombinant ANGPTL1/Angiopoietin-like 1 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the canine ANGPTL1 (E2RQY3) (Glu197-Asp491) was expressed with the Fc region of human IgG1 at the N-terminus.