Mouse Recombinant OX-40L/TNFSF4/CD252 Protein (rFc Tag) (from HEK293 Cells)
Supplier: Sino Biological
53582-M15H-50
103668-836EA 179.78 USD
103668-836
Mouse Recombinant OX-40L/TNFSF4/CD252 Protein (rFc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse Tnfsf4 ( NP_033478.1) (Gln49-Leu198) was expressed with the Fc region of rabbit IgG at the N-terminus..