Mouse Recombinant ANTXR2/CMG2 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
51131-M08H-100 51131-M08H-50
103665-830EA 300.98 USD
103665-830 103668-600
Mouse Recombinant ANTXR2/CMG2 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of mouse ANTXR2 (NP_598499.1) (Met1-Gly318) was expressed with a C-terminal polyhistidine tag.