Mouse Recombinant FLRT2 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
51074-M08H-20 51074-M08H-50
103668-488EA 179.78 USD
103668-488 103668-430
Mouse Recombinant FLRT2 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse FLRT2 (Q8BLU0) (Met1-Ser539) was expressed with a C-terminal polyhistidine tag.