Mouse Recombinant HER2/ErbB2 Protein (ECD, Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
50714-M02H-100
103665-418EA 300.98 USD
103665-418
Mouse Recombinant HER2/ErbB2 Protein (ECD, Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse ERBB2 (P70424) extracellular domain (Met 1-Thr 653) was fused with the Fc region of human IgG1 at the C-terminus.