Mouse Recombinant Semaphorin 3A/SEMA3A Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
50631-M01H-20 50631-M01H-50
103667-774EA 116.1 USD
103667-774 103667-776
Mouse Recombinant Semaphorin 3A/SEMA3A Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the N-terminal fragment (Lys 26-Phe 546) of mouse SEMA3A (O08665) was fused with the Fc region of human IgG1 at the N-terminus.