Mouse Recombinant Peroxiredoxin 5/PRDX5 Protein (His Tag) (from E. coli)
Supplier: Sino Biological
50551-M07E-100 50551-M07E-50
103665-140EA 250.48 USD
103665-140 103667-670
Mouse Recombinant Peroxiredoxin 5/PRDX5 Protein (His Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the mouse PRDX5 (mitochondrial) (P99029-1) (Met 49-Leu 210) was expressed, with a polyhistide tag at the N-terminus.