Mouse Recombinant BMP4/BMP-4 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
50439-M15H-50 50439-M15H-100 50439-M15H-5 50439-M15H-10
103664-990EA 452.48 USD
103664-990 103677-828 103667-440 103667-442
Mouse Recombinant BMP4/BMP-4 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse BMP4 (NP_031580.2) (Ser293-Arg408) was expressed with the Fc region of rabbit IgG at the N-terminus..