Mouse Recombinant SerpinA3c Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
50375-M08H-50 50375-M08H-100
103667-318EA 191.35 USD
103667-318 103664-936
Mouse Recombinant SerpinA3c Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of mouse SERPINA3C (NP_032484.1) precursor (Met 1-Ala 417) was expressed, with a C-terminal polyhistidine tag.