Mouse Recombinant Coagulation Factor IX/FIX/F9 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
50362-M08H-20 50362-M08H-10
103667-296EA 148.35 USD
103667-296 103667-298
Mouse Recombinant Coagulation Factor IX/FIX/F9 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the full length of mouse F9 (P16294) (Met 1-Thr 471) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.